Assay Details
Gene Symbol: CUNH11orf95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852567.3 | XP_004852624.1 | 4 - 5 | 1536 | 100 | |
XM_021264703.1 | XP_021120362.1 | 4 - 5 | 3184 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
