Assay Details
| Gene Symbol: Snrnp70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004866960.3 | XP_004867017.1 | 5 - 6 | 863 | 64 | |
| XM_013066336.2 | XP_012921790.1 | 5 - 6 | 864 | 64 | ||
| XM_021246168.1 | XP_021101827.1 | 5 - 6 | 457 | 64 | ||
More Information
Set Membership: |
|

Snrnp70
Genomic Map