Assay Details
Gene Symbol: LOC101712173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873124.3 | XP_004873181.1 | 4 - 5 | 1412 | 70 | |
XM_021250889.1 | XP_021106548.1 | 3 - 4 | 1235 | 70 |

More Information
Set Membership: |
![]() ![]() |
