Assay Details
Gene Symbol: LOC101712173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004873124.3 | XP_004873181.1 | 6 - 7 | 1682 | 101 | |
XM_021250889.1 | XP_021106548.1 | 5 - 6 | 1505 | 101 |

More Information
Set Membership: |
![]() ![]() |
