Assay Details
Gene Symbol: Pkmyt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013065654.2 | XP_012921108.1 | 4 - 5 | 1270 | 63 | |
XM_013065655.2 | XP_012921109.1 | 3 - 4 | 1125 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
