Assay Details
Gene Symbol: Ddx39a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068378.2 | XP_012923832.1 | 8 - 9 | 1078 | 64 | |
XM_021250413.1 | XP_021106072.1 | 8 - 9 | 1179 | 64 | ||
XM_021250414.1 | XP_021106073.1 | 8 - 9 | 1154 | 64 | ||
XM_021250415.1 | XP_021106074.1 | 7 - 8 | 1040 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
