Assay Details
Gene Symbol: LOC101723821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013073804.2 | XP_012929258.1 | 13 - 14 | 1985 | 70 | |
XM_013073806.2 | XP_012929260.1 | 12 - 13 | 1800 | 70 | ||
XM_013073807.2 | XP_012929261.1 | 11 - 12 | 1602 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
