Assay Details
| Gene Symbol: LOC101723821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073804.2 | XP_012929258.1 | 15 - 16 | 2272 | 108 | |
| XM_013073805.2 | XP_012929259.1 | 14 - 15 | 2215 | 108 | ||
| XM_013073806.2 | XP_012929260.1 | 14 - 15 | 2087 | 108 | ||
| XM_013073807.2 | XP_012929261.1 | 13 - 14 | 1889 | 108 | ||
More Information
Set Membership: |
|

LOC101723821
Genomic Map