Assay Details
| Gene Symbol: LOC101723821 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073804.2 | XP_012929258.1 | 1 - 2 | 318 | 106 | |
| XM_013073805.2 | XP_012929259.1 | 1 - 2 | 324 | 106 | ||
| XM_013073806.2 | XP_012929260.1 | 1 - 2 | 319 | 106 | ||
| XM_013073807.2 | XP_012929261.1 | 1 - 2 | 325 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013073804.2, XM_013073805.2, XM_013073806.2, XM_013073807.2 transcripts.
Set Membership: |
|

LOC101723821
Genomic Map