Assay Details
Gene Symbol: LOC101723821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013073804.2 | XP_012929258.1 | 6 - 7 | 931 | 89 | |
XM_013073805.2 | XP_012929259.1 | 6 - 7 | 937 | 89 | ||
XM_013073806.2 | XP_012929260.1 | 6 - 7 | 932 | 89 | ||
XM_013073807.2 | XP_012929261.1 | 6 - 7 | 938 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
