Assay Details
| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248908.1 | XP_021104567.1 | 1 - 2 | 262 | 94 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rasl10b| Gene Symbol: Rasl10b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248908.1 | XP_021104567.1 | 1 - 2 | 262 | 94 | |