Assay Details
Gene Symbol: Mxra7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860766.3 | XP_004860823.1 | 2 - 3 | 395 | 110 | |
XM_004860767.2 | XP_004860824.1 | 2 - 3 | 392 | 110 | ||
XM_004860769.2 | XP_004860826.1 | 2 - 3 | 392 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
