Assay Details
| Gene Symbol: Dohh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865794.2 | XP_004865851.1 | 2 - 3 | 606 | 82 | |
| XM_004865795.2 | XP_004865852.1 | 2 - 3 | 625 | 82 | ||
| XM_013065859.2 | XP_012921313.1 | 2 - 3 | 560 | 82 | ||
More Information
Set Membership: |
|

Dohh
Genomic Map