Assay Details
Gene Symbol: Ca11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866987.3 | XP_004867044.2 | 6 - 7 | 850 | 63 | |
XM_004866988.3 | XP_004867045.1 | 7 - 8 | 1093 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
