Assay Details
Gene Symbol: Creb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870139.3 | XP_004870196.1 | 3 - 4 | 394 | 64 | |
XM_013067555.2 | XP_012923009.1 | 3 - 4 | 409 | 64 | ||
XM_013067557.2 | XP_012923011.1 | 3 - 4 | 411 | 64 | ||
XM_021248517.1 | XP_021104176.1 | 3 - 4 | 413 | 64 | ||
XM_021248518.1 | XP_021104177.1 | 3 - 4 | 575 | 64 | ||
XM_021248519.1 | XP_021104178.1 | 3 - 4 | 416 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
