Assay Details
Gene Symbol: Pqlc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004921035.1 | XP_004921092.1 | 2 - 3 | 256 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pqlc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004921035.1 | XP_004921092.1 | 2 - 3 | 256 | 80 |
Set Membership: |
![]() ![]() ![]() ![]() |