Assay Details
Gene Symbol: LOC101717066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072797.2 | XP_012928251.1 | 4 - 5 | 1640 | 86 | |
XM_021260072.1 | XP_021115731.1 | 3 - 4 | 552 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
