Assay Details
Gene Symbol: LOC101717066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072797.2 | XP_012928251.1 | 7 - 8 | 1990 | 91 | |
XM_021260072.1 | XP_021115731.1 | 6 - 7 | 902 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
