Assay Details
Gene Symbol: LOC101717066 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072797.2 | XP_012928251.1 | 9 - 10 | 2273 | 66 | |
XM_021260072.1 | XP_021115731.1 | 8 - 9 | 1185 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
