Assay Details
Gene Symbol: LOC101716216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021242809.1 | XP_021098468.1 | 18 - 19 | 1727 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
