Assay Details
Gene Symbol: Rnase10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021244540.1 | XP_021100199.1 | 2 - 3 | 115 | 69 | |
XM_021244541.1 | XP_021100200.1 | 2 - 3 | 167 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021244540.1, XM_021244541.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
