Assay Details
Gene Symbol: Bhmg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021246181.1 | XP_021101840.1 | 10 - 11 | 1361 | 87 | |
XM_021246183.1 | XP_021101842.1 | 10 - 11 | 1358 | 87 | ||
XM_021246184.1 | XP_021101843.1 | 10 - 11 | 1286 | 87 |
