Assay Details
Gene Symbol: Glipr1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021258317.1 | XP_021113976.1 | 1 - 2 | 58 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
