Assay Details
Gene Symbol: Shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835475.2 | XP_004835532.2 | 2 - 3 | 629 | 54 | |
XM_013074552.1 | XP_012930006.1 | 2 - 3 | 629 | 54 | ||
XM_013074556.2 | XP_012930010.1 | 2 - 3 | 629 | 54 | ||
XM_013074557.2 | XP_012930011.1 | 2 - 3 | 629 | 54 | ||
XM_021261659.1 | XP_021117318.1 | 2 - 3 | 629 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
