Assay Details
Gene Symbol: Snurf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004852639.3 | XP_004852696.1 | 2 - 3 | 299 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004852639.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
