Assay Details
Gene Symbol: Gp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855639.2 | XP_004855696.1 | 1 - 2 | 527 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004855639.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
