Assay Details
Gene Symbol: Bglap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871189.3 | XP_004871246.1 | 1 - 2 | 979 | 115 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004871189.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
