Assay Details
| Gene Symbol: Dmrtc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004873071.2 | XP_004873128.1 | 5 - 6 | 849 | 56 | |
| XM_004873072.2 | XP_004873129.1 | 5 - 6 | 769 | 56 | ||
| XM_004873074.2 | XP_004873131.1 | 5 - 6 | 849 | 56 | ||
| XM_021250970.1 | XP_021106629.1 | 5 - 6 | 802 | 56 | ||
More Information
Set Membership: |
|

Dmrtc2
Genomic Map