Assay Details
Gene Symbol: Sprr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013067968.2 | XP_012923422.1 | 1 - 2 | 79 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013067968.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
