Assay Details
| Gene Symbol: Ripk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073264.2 | XP_012928718.1 | 8 - 9 | 1306 | 73 | |
| XM_021260742.1 | XP_021116401.1 | 8 - 9 | 1314 | 73 | ||
More Information
Set Membership: |
|

Ripk1
Genomic Map