Assay Details
Gene Symbol: Tnfsf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013076789.2 | XP_012932243.1 | 5 - 6 | 853 | 88 | |
XM_013076790.2 | XP_012932244.1 | 5 - 6 | 850 | 88 | ||
XM_021238148.1 | XP_021093807.1 | 5 - 6 | 465 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
