Assay Details
Gene Symbol: LOC101703392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834207.3 | XP_004834264.1 | 1 - 2 | 164 | 111 | |
XM_021241509.1 | XP_021097168.1 | 1 - 2 | 183 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004834207.3, XM_021241509.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
