Assay Details
Gene Symbol: LOC101703392 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834207.3 | XP_004834264.1 | 4 - 5 | 517 | 84 | |
XM_021241509.1 | XP_021097168.1 | 4 - 5 | 536 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
