Assay Details
| Gene Symbol: LOC101703392 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834207.3 | XP_004834264.1 | 7 - 8 | 898 | 67 | |
| XM_021241509.1 | XP_021097168.1 | 7 - 8 | 917 | 67 | ||
More Information
Set Membership: |
|

LOC101703392
Genomic Map