Assay Details
| Gene Symbol: Dnaaf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835585.3 | XP_004835642.1 | 1 - 2 | 1772 | 78 | |
| XM_021262610.1 | XP_021118269.1 | 1 - 2 | 1772 | 78 | ||
More Information
Set Membership: |
|

Dnaaf2
Genomic Map