Assay Details
Gene Symbol: Avpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838610.2 | XP_004838667.1 | 1 - 2 | 199 | 104 | |
XM_013069913.2 | XP_012925367.1 | 1 - 2 | 199 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004838610.2, XM_013069913.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
