Assay Details
Gene Symbol: Apcdd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841166.3 | XP_004841223.1 | 2 - 3 | 979 | 72 | |
XM_021255163.1 | XP_021110822.1 | 2 - 3 | 262 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
