Assay Details
Gene Symbol: Apcdd1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841166.3 | XP_004841223.1 | 3 - 4 | 1532 | 97 | |
XM_021255163.1 | XP_021110822.1 | 3 - 4 | 815 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
