Assay Details
| Gene Symbol: Ckmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841622.3 | XP_004841679.1 | 7 - 8 | 1106 | 61 | |
| XM_013071088.2 | XP_012926542.1 | 8 - 9 | 1265 | 61 | ||
| XM_021255712.1 | XP_021111371.1 | 7 - 8 | 1085 | 61 | ||
| XM_021255713.1 | XP_021111372.1 | 8 - 9 | 3129 | 61 | ||
| XM_021255714.1 | XP_021111373.1 | 8 - 9 | 2292 | 61 | ||
More Information
Set Membership: |
|

Ckmt2
Genomic Map