Assay Details
| Gene Symbol: Cabp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843412.3 | XP_004843469.1 | 3 - 4 | 381 | 72 | |
| XM_004843413.3 | XP_004843470.2 | 3 - 4 | 381 | 72 | ||
| XM_004843414.3 | XP_004843471.2 | 3 - 4 | 381 | 72 | ||
More Information
Set Membership: |
|

Cabp7
Genomic Map