Assay Details
Gene Symbol: CUNH6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846669.2 | XP_004846726.1 | 2 - 3 | 88 | 82 | |
XM_004846671.2 | XP_004846728.1 | 2 - 3 | 246 | 82 | ||
XM_013072804.2 | XP_012928258.1 | 2 - 3 | 184 | 82 | ||
XM_013072805.2 | XP_012928259.1 | 2 - 3 | 177 | 82 | ||
XM_021259904.1 | XP_021115563.1 | 2 - 3 | 177 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004846669.2, XM_004846671.2, XM_013072804.2, XM_013072805.2, XM_021259904.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
