Assay Details
Gene Symbol: CUNH6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846669.2 | XP_004846726.1 | 5 - 6 | 669 | 85 | |
XM_004846671.2 | XP_004846728.1 | 5 - 6 | 827 | 85 | ||
XM_013072804.2 | XP_012928258.1 | 5 - 6 | 765 | 85 | ||
XM_013072805.2 | XP_012928259.1 | 5 - 6 | 758 | 85 | ||
XM_021259904.1 | XP_021115563.1 | 5 - 6 | 758 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
