Assay Details
Gene Symbol: CUNH6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846669.2 | XP_004846726.1 | 6 - 7 | 809 | 102 | |
XM_004846671.2 | XP_004846728.1 | 6 - 7 | 967 | 102 | ||
XM_013072804.2 | XP_012928258.1 | 6 - 7 | 905 | 102 | ||
XM_013072805.2 | XP_012928259.1 | 6 - 7 | 898 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
