Assay Details
Gene Symbol: CUNH6orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846669.2 | XP_004846726.1 | 7 - 8 | 929 | 103 | |
XM_004846671.2 | XP_004846728.1 | 7 - 8 | 1087 | 103 | ||
XM_013072804.2 | XP_012928258.1 | 7 - 8 | 1025 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
