Assay Details
| Gene Symbol: CUNH6orf89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846669.2 | XP_004846726.1 | 8 - 9 | 1063 | 108 | |
| XM_004846671.2 | XP_004846728.1 | 8 - 9 | 1221 | 108 | ||
| XM_013072804.2 | XP_012928258.1 | 8 - 9 | 1159 | 108 | ||
More Information
Set Membership: |
|

CUNH6orf89
Genomic Map