Assay Details
Gene Symbol: Hfe2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853927.2 | XP_004853984.1 | 3 - 4 | 1219 | 105 | |
XM_004853928.2 | XP_004853985.1 | 3 - 4 | 936 | 105 | ||
XM_021265830.1 | XP_021121489.1 | 3 - 4 | 1309 | 105 | ||
XM_021265832.1 | XP_021121491.1 | 4 - 5 | 1115 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
