Assay Details
| Gene Symbol: Aldoa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856093.3 | XP_004856150.1 | 4 - 5 | 575 | 60 | |
| XM_004856094.3 | XP_004856151.1 | 4 - 5 | 484 | 60 | ||
| XM_004856095.3 | XP_004856152.1 | 4 - 5 | 466 | 60 | ||
| XM_013076292.2 | XP_012931746.1 | 5 - 6 | 674 | 60 | ||
More Information
Set Membership: |
|

Aldoa
Genomic Map