Assay Details
| Gene Symbol: Dedd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004858682.3 | XP_004858739.1 | 2 - 3 | 1804 | 65 | |
| XM_013077499.2 | XP_012932953.1 | 2 - 3 | 555 | 65 | ||
| XM_013077500.2 | XP_012932954.1 | 3 - 4 | 520 | 65 | ||
| XM_013077501.2 | XP_012932955.1 | 3 - 4 | 588 | 65 | ||
| XM_013077502.2 | XP_012932956.1 | 2 - 3 | 481 | 65 | ||
| XM_021239614.1 | XP_021095273.1 | 3 - 4 | 1490 | 65 | ||
| XM_021239615.1 | XP_021095274.1 | 3 - 4 | 543 | 65 | ||
More Information
Set Membership: |
|

Dedd
Genomic Map