Assay Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004863000.2 | XP_004863057.1 | 4 - 5 | 474 | 60 | |
XM_004863001.2 | XP_004863058.1 | 3 - 4 | 413 | 60 | ||
XM_004863002.2 | XP_004863059.1 | 3 - 4 | 390 | 60 | ||
XM_013079085.2 | XP_012934539.1 | 4 - 5 | 470 | 60 | ||
XM_021243112.1 | XP_021098771.1 | 4 - 5 | 451 | 60 | ||
XM_021243113.1 | XP_021098772.1 | 4 - 5 | 350 | 60 | ||
XM_021243114.1 | XP_021098773.1 | 4 - 5 | 418 | 60 | ||
XM_021243115.1 | XP_021098774.1 | 4 - 5 | 433 | 60 | ||
XM_021243116.1 | XP_021098775.1 | 2 - 3 | 304 | 60 | ||
XM_021243117.1 | XP_021098776.1 | 5 - 6 | 488 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
