Assay Details
Gene Symbol: LOC101703228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865960.3 | XP_004866017.1 | 2 - 3 | 687 | 75 | |
XM_013065931.2 | XP_012921385.1 | 2 - 3 | 688 | 75 | ||
XM_013065933.2 | XP_012921387.1 | 2 - 3 | 221 | 75 | ||
XM_021245171.1 | XP_021100830.1 | 2 - 3 | 688 | 75 | ||
XM_021245172.1 | XP_021100831.1 | 2 - 3 | 689 | 75 | ||
XM_021245173.1 | XP_021100832.1 | 1 - 2 | 115 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021245173.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
